Learn More
Idex Valves
Easily switch between two fluid paths without disconnecting the lines
Manufacturer:  Idex V100T
 View more versions of this product
Catalog No. 05-704-20
Add to cart
Description
Description
Easily switch between two fluid paths without disconnecting the lines.
  • Enable connections among multiple fluid systems
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.
Specifications
Specifications
1.80 in.
0.04 in.
500 psi
0.66 in.
9.7 µL