Learn More
Idex VHP Union Assembly
Zero dead volume unions ensure accurate analysis
Manufacturer:  Idex U402
 View more versions of this product
Catalog No. 05-700-231
Add to cart
Description
Description
  • Precision machined from 316 stainless steel to withstand high pressures
  • Securely join fluid pathway components Holds tight connection even through vibration
  • VHP ZDV union assembly
  • 10-32 coned for 1/16 in. OD tubing
  • Max pressure 20,000 psi (1380 bar)
Specifications
Specifications
Stainless Steel
Union
0.5 mm
0.134 µL
1/16 in. O.D. Tubing
0.02 in.
10000 psi