Learn More
UPF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233791AP
Catalog No. 50-173-4064
Add to cart
Description
Description
Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
Specifications
Specifications
UPF1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92900, Q9EPU0
ATP dependent helicase RENT1, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1
Rabbit
IgG
150 μL
-20°C
Primary
19704, 5976, 684558
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
UPF1
Liquid
UPF1
UPF1 Fusion Protein Ag16195
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat