Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPK1A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549668

Catalog No. PA549668

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.
Specifications

Specifications

UPK1A
Polyclonal
Unconjugated
UPK1A
Liquid
UPK1A
KLH conjugated synthetic peptide between 191-217 amino acids from the C-terminal region of human UPK1A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide
O00322
Uroplakin-1a, UP1a, Tetraspanin-21, Tspan-21, Uroplakin Ia, UPIa, UPKa, UPK1A, TSPAN21
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11045
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit