Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UPK3B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548890
Description
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence.Specifications
UPK3B | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9BT76 | |
UPK3B | |
KLH conjugated synthetic peptide between 192-219 amino acids from the Central region of human UPK3B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UPK3B | |
Uroplakin-3b, UP3b, Uroplakin IIIb, UPIIIb, p35, UPK3B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105375355 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction