Learn More
UPK3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157091AP
Catalog No. 50-173-0422
Add to cart
Description
Description
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence.
Specifications
Specifications
UPK3B
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q80YF6, Q9BT76
P35, UP3b, UPIIIb, UPK3B, uroplakin 3B, Uroplakin IIIb
Rabbit
IgG
150 μL
-20°C
Primary
100647, 105375355
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
UPK3B
Liquid
UPK3B
UPK3B Fusion Protein Ag8326
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse