Learn More
Meso Scale Discovery UPLEX Mouse KC/GRO ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K152VWK4
Encompass
The U-PLEX Mouse KC Assay K152VWK from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantification of 15 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A/F IFN- TNF- GM-CSF KC/GRO MCP-1 MIP-1 ) from cell culture supernatants serum plasma and other biological samples This 10-plate kit utilizes MSD s electrochemiluminescent detection technology for high sensitivity wide dynamic range and minimal matrix interference The assay requires 50-100 L of sample per well and can be read on MSD s SECTOR Imager instrumentation
Catalog No. 50-000-79350
Add to Cart
Request a Quote