Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550894

Catalog No. PA550894


Add to cart

Description

Description

The antibody detects endogenous levels of total UPP2 protein.

Catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate. The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis. Shows substrate specificity and accept uridine, deoxyuridine, and thymidine as well as the two pyrimidine nucleoside analogs 5-fluorouridine and 5-fluoro-2( ft.)-deoxyuridine as substrates.
Specifications

Specifications

UPP2
Polyclonal
Unconjugated
UPP2
Liquid
UPP2
Synthetic peptide of human uridine phosphorylase 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
5.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O95045, Q8CGR7
UP2, UPASE2, UDRPASE2
Rabbit
IgG
100 μL
-20°C
Primary
151531, 76654
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit