Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRB Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548493
Description
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This component is involved in redox-linked proton pumping.Specifications
UQCRB | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P14927 | |
UQCRB | |
KLH conjugated synthetic peptide between 19-47 amino acids from the Central region of human UQCRB. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
UQCRB | |
Cytochrome b-c1 complex subunit 7, Complex III subunit 7, Complex III subunit VII, QP-C, Ubiquinol-cytochrome c reductase complex 14 kDa protein, UQCRB, UQBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7381 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction