Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UQCRC1 Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: 139, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243597G

Catalog No. NBP243597G


Add to cart

Description

Description

UQCRC1 Monoclonal antibody specifically detects UQCRC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

UQCRC1
139
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
UQCRC1
Recombinant protein encompassing a sequence within the center region of human UQCRC1. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Complex III subunit 1, Core protein I, cytochrome b-c1 complex subunit 1, mitochondrial, D3S3191, QCR1, ubiquinol-cytochrome c reductase core protein I, Ubiquinol-cytochrome-c reductase complex core protein 1, UQCR1
Mouse
IgG1
0.1 ml
Primary
7384
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit