Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UQCRH Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551310

Catalog No. PA551310


Add to cart

Description

Description

The antibody detects endogenous levels of total UQCRH protein.

Cytochrome b-c1 complex subunit 6, mitochondrial is a protein that in humans is encoded by the UQCRHgene. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
Specifications

Specifications

UQCRH
Polyclonal
Unconjugated
UQCRH
Liquid
UQCRH
Full length fusion protein of human UQCRH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P07919, P99028
QCR6, UQCR8
Rabbit
IgG
100 μL
-20°C
Primary
66576, 7388
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit