Alfa Aesar™ Uracil-5-boronic acid, 95%

Manufacturer: Alfa Aesar™ H27219MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27219MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70523-22-7
    Molecular Formula C4H5BN2O4
    Formula Weight 155.90g/mol
    MDL Number MFCD01318983
    Synonym 2,4-Dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid
    Chemical Name or Material Uracil-5-boronic acid

    Specifications

    Melting Point >300°C
    Quantity 250mg
    CAS 70523-22-7
    Molecular Formula C4H5BN2O4
    Formula Weight 155.90g/mol
    MDL Number MFCD01318983
    Synonym 2,4-Dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    Chemical Name or Material Uracil-5-boronic acid
    Assay 95%

    Description

    Applications
    It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    −30° to −10°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit