Alfa Aesar™ Uracil-6-carboxaldehyde monohydrate, 98%

Manufacturer: Alfa Aesar™ H50469MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50469MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36327-91-0
    Molecular Formula C5H4N2O3·H2O
    Formula Weight 158.11 (140.10 Anhydrous)
    MDL Number MFCD03002359
    Synonym 6-Formyluracil monohydrate
    Chemical Name or Material Uracil-6-carboxaldehyde monohydrate

    Specifications

    Formula Weight 158.11 (140.10 Anhydrous)
    Molecular Formula C5H4N2O3·H2O
    MDL Number MFCD03002359
    CAS 36327-91-0
    Synonym 6-Formyluracil monohydrate
    Quantity 250mg
    Chemical Name or Material Uracil-6-carboxaldehyde monohydrate
    Assay 98%

    Description

    Applications
    Uracil-6-carboxaldehyde monohydrate is used as pharmaceutical intermediates.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit