Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URB1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550077

Catalog No. PA550077


Add to cart

Description

Description

The antibody detects endogenous levels of total URB1 protein.

The German cDNA consortium; Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases. Slavov D., Gene 247:215-232(2000). Nagase T., DNA Res. 5:31-39(1998).
Specifications

Specifications

URB1
Polyclonal
Unconjugated
URB1
Liquid
URB1
Synthesized peptide derived from C-terminal of human URB1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O60287
Nucleolar pre-ribosomal-associated protein 1, Protein URB1,
Rabbit
IgG
100 μL
-20°C
Primary
9875
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit