Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Urea, 99.0 to 100.5%, Thermo Scientific™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
57-13-6
CH4N2O
60.056
MFCD00008022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
57-13-6
CH4N2O
60.056
MFCD00008022
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
Specific Gravity 1.335
Packaging Plastic bottle
Quantity 1kg
Melting Point 132°C to 135°C
Formula Weight 60.06
Physical Form Crystalline Powder
Percent Purity 99.0 to 100.5%
Chemical Name or Material Urea