Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Plastic bag in resealable cardbox
Plastic bottle
Plastic drum
57-13-6
CH4N2O
60.06
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
57-13-6
CH4N2O
60.06
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
For analysis
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 131.0°C to 135.0°C
Quantity 1kg
Color White
Assay Percent Range 99.5%
Linear Formula H2NCONH2
Beilstein 03,42
Fieser 01,1262; 02,455
Merck Index 15,10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol,benzene,concentrated hcl,practically insoluble in chloroform and ether,soluble in methanol: 16.6g/100 mL,soluble in ethanol: 5g/100 mL
Formula Weight 60.06
Physical Form Fine Crystals
Percent Purity 99.5%
Grade Analysis
Chemical Name or Material Urea
Show More Show Less