Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Description
- Chaotropic agent used in denaturing polyacrylamide gel electrophoresis.
- When 8M urea is not sufficiently denaturing, 40% formamide may be included.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 132°C to 135°C |
Density | 1.335 |
Packaging | Plastic bottle |
Linear Formula | NH2CONH2 |
Quantity | 1 kg |
Beilstein | 635724 |
Specific Gravity | 1.335 |
Formula Weight | 60.06 |
Percent Purity | ≥99.5% |
Grade | Molecular Biology |
Show More |
Safety and Handling
EINECSNumber | 200-315-5 |
RTECSNumber | YR6250000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction