Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N
57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate
1176
CHEBI:48376
urea
C(=O)(N)N

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Specific Gravity 1L = 1.32kg
Packaging Poly Bottle
Formula Weight 60.06
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Urea