Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, cryst. EMPROVE™ EXPERT Ph Eur, BP, JP, USP, ACS, 102.0%, MilliporeSigma™

Click to view available options
Quantity:
12 kg
CAS 57-13-6
Density 1.34 g/cm3 (20°C)
pH 9 (100 g/L, H2O, 20°C)
Melting Point 113°C
Assay Percent Range 98.0 to 102.0 % by HPLC
Molecular Formula CO(NH2)2
Packaging Plastic Bucket
Quantity 12 kg
Solubility Information 1000 g/L
Molecular Weight (g/mol) 60.06
Grade Ph Eur, BP, JP, USP, ACS
Vapor Pressure < 0.1 hPa (20°C)
Show More Show Less

Recommended Storage : Store at 25 to 30°C