Learn More
Thermo Scientific™ Urea Reagents
Determine quantity of urea in serum, plasma or urine in vitro with Thermo Scientific™ Urea Reagents.
Catalog No. TR12421
Add to cart
Description
Description
Determine quantity of urea in serum, plasma or urine in vitro with Thermo Scientific™ Urea Reagents, suitable for manual use or on automated clinical chemistry analyzers.
Urea is the major end product of protein nitrogen metabolism. Elevated levels can occur with dietary changes, diseases that impair kidney function, liver disease, congestive heart failure, diabetes and infection.
  • Offers a 3-month reconstituted stability and various kit sizes to minimize reagent wastage
  • Applications for a wide range of automated analyzers available on request
Specifications
Specifications
CE Marked
Plasma, Serum, Urine
Reagent
Infinity BUN / Urea
0.5-40mmol/L (3-112mg/dL)
2 x 125 mL
Liquid
2°C to 8°C
Yes
Urea
Fixed rate
Until expiry at 2-8°C
SDS
Documents
Accessories