Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, Shotted Pellets, Reagents

Catalog No. 01336898
Click to view available options
Quantity:
500 g
2.5 kg

Urea, Shotted Pellets, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O

Specifications

Boiling Range 135°C
Melting Point 133°C
Color White
Boiling Point 135°C
Melting Range 133°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1.335
Grade Technical
Physical Form Solid
Chemical Name or Material Urea, Shotted Pellets
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.