Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urea, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

57-13-6
CH4N2O
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
CHEBI:48376
urea
C(=O)(N)N
Packaging Poly Bottle
Specific Gravity 1L = 1.32kg
Formula Weight 60.06
Quantity 500g
Chemical Name or Material Urea
Grade USP