Learn More
Sklar InstrumentsSupplier Diversity Partner Guyon Urethral Sound without Thread, Sklar, 10-1/4 in., Stainless Steel, 34 French. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 852534
Guyon Urethral Sound without Thread, Sklar, 10-1/4 in., Stainless Steel, 34 French. Sklar's Guyon Urethral Sound is used to increase the inner diameter of the urethra to locate obstructions or for dilation prior to endoscopic procedures. The instrument has a straight shaft with a large curve leading into the tip. This instrument has a 34 French (11.3mm) tip.
Catalog No. 50-143-6465