Learn More
Sklar InstrumentsSupplier Diversity Partner Guyon Urethral Sound with Thread, Sklar, 10-3/4 in., 28 French, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 852628
Guyon Urethral Sound with Thread, Sklar, 10-3/4 in., 28 French, Stainless Steel. Sklar's Guyon Urethral Sound with Thread is used with standard 1-56 thread for filiform to insert through the urethra. The filiform is attached to the end of the probe. The flat handle provides improved grip and greater control. This product is a size 28 French (9.3mm) curved with an overall length of 10-3/4 inches.
Catalog No. 50-143-6494