Learn More
Aviva Systems Biology Corp URG4 Antibody - middle region (ARP56288_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56288_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against URG4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human URG4. Alternate names: DKFZp666G166, DKFZp686O0457, URG4ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-61796