Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URG4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549670

Catalog No. PA549670

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in cell cycle progression through the regulation of cyclin D1 expression. May participate in the development of hepatocellular carcinoma (HCC) by promoting hepatocellular growth and survival. May play an important role in development of gastric cancer.
Specifications

Specifications

URG4
Polyclonal
PBS with 0.09% sodium azide
Q8TCY9
Up-regulator of cell proliferation, HBV X protein up-regulated gene 4 protein, HBxAg up-regulated gene 4 protein, URGCP, KIAA1507, URG4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55665
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
URGCP
Liquid
URGCP
KLH conjugated synthetic peptide between 466-495 amino acids from the Central region of human URG4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit