Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uric acid, +99%, ACROS Organics™

Click to view available options
Quantity:
1kg
Packaging:
Plastic bottle
69-93-2
C5H4N4O3
168.112
MFCD00005712
LEHOTFFKMJEONL-UHFFFAOYSA-N
uric acid,urate,2,6,8-trioxypurine,lithic acid,2,6,8-trihydroxypurine,2,6,8-trioxopurine,1h-purine-2,6,8-triol,8-hydroxyxanthine,7,9-dihydro-1h-purine-2,6,8 3h-trione,1h-purine-2,6,8 3h-trione, 7,9-dihydro
1175
CHEBI:17775
7,9-dihydro-3H-purine-2,6,8-trione
C12=C(NC(=O)N1)NC(=O)NC2=O
69-93-2
C5H4N4O3
168.112
MFCD00005712
LEHOTFFKMJEONL-UHFFFAOYSA-N
uric acid,urate,2,6,8-trioxypurine,lithic acid,2,6,8-trihydroxypurine,2,6,8-trioxopurine,1h-purine-2,6,8-triol,8-hydroxyxanthine,7,9-dihydro-1h-purine-2,6,8 3h-trione,1h-purine-2,6,8 3h-trione, 7,9-dihydro
1175
CHEBI:17775
7,9-dihydro-3H-purine-2,6,8-trione
C12=C(NC(=O)N1)NC(=O)NC2=O
Quantity 1kg
Infrared Spectrum Authentic
Color White
Sulfated Ash 0.2% max.
Loss on Drying 0.5% max. (105°C, 3 hrs)
Packaging Plastic bottle
Assay Percent Range 99+%
Formula Weight 168.11
Physical Form Fine Powder
Percent Purity ≥99.0% (ex N)
Chemical Name or Material Uric acid, 99+%
Show More Show Less

EINECSNumber : 200-720-7

RTECSNumber : YU7050080

TSCA : TSCA