Learn More
Thermo Scientific Uric Acid Reagents
Determine quantity of uric acid in human serum, plasma or urine in vitro with Thermo Scientific™ Uric Acid Reagents.
Description
Determine quantity of uric acid in human serum, plasma or urine in vitro with Thermo Scientific Uric Acid Reagents, offering up to 18 months of stability from the manufacture date.
Uric acid is a metabolite of purines, nucleic acids and nucleoproteins. Hyperuricaemia may be observed in renal dysfunction, gout, leukemia, toxemia of pregnancy or some genetic disorders. Decreased levels may be observed with severe hepatocellular disease and defective renal tubular reabsorption.Specifications
CE Marked | |
Reagent | |
Infinity Uric Acid Liquid Stable Reagent | |
0.03-1.5mmol/L (0.5-25.2mg/dL) | |
Until expiry at 2-8°C |
Plasma, Serum, Serum or Urine | |
Yes | |
Uric Acid | |
Enzymatic endpoint |