Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copan Diagnostics™ UriSponge™: Urine Specimen Collection, Preservation and Transport System

Allows safe collection and preservation of urine specimens without the added expense and hazard of hypodermic urine transfer straws
Specifications
Product Type | Foam Sponge Urine Collection Device |
---|---|
Closure Type | Screw Cap |
Material | Polypropylene with Polyurethane Foam Sponge |
Catalog Number | Mfr. No. | Tube Size | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Tube Size | Packaging | Price | Quantity & Availability | ||||
23-600-529
![]() |
Copan Diagnostics™
8C023S01.A |
12 x 80 mm | Individual Peel Pouch | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-600-531
![]() |
Copan Diagnostics™
8C022S100.A |
12 x 80 mm | Bulk | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-600-527
![]() |
Copan Diagnostics™
8C021S01.A |
16 x 100 mm | Individual Peel Pouch | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
23-600-528
![]() |
Copan Diagnostics™
8C020S100.A |
16 x 100 mm | Bulk | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Urine collection device combines a plastic tube and a screw cap with attached sponges impregnated with boric acid and sodium formate urine preservatives
- Maintains viability of bacteria at refrigerated and room temperature for up to 48 hours
- No hypodermic needle urine transfer straw needed, eliminating potential injury and sharps disposal costs
- Reduce repeat collections from patient due to inaccurate filling of boric acid tubes
- Automatically processed on WASP™ using its built-in spinner, which easily elutes urine from the sponges prior to planting and streaking
- If processed manually, simply centrifuge the tube to release urine from the sponge
Specifications
Foam Sponge Urine Collection Device | |
Screw Cap | |
Polypropylene with Polyurethane Foam Sponge |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title