Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UROC1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548130
Description
This gene encodes an enzyme involved in histidine catabolism, metabolizing urocanic acid to formiminoglutamic acid. The gene product is known to protect the skin from ultra violet rays and is contained in human sweat. Deficiency of this gene product in the liver is an apparent cause of mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Sep 2009]Specifications
UROC1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96N76 | |
UROC1 | |
KLH conjugated synthetic peptide between 402-430 amino acids from the Central region of human UROC1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
UROC1 | |
Urocanate hydratase, Urocanase, Imidazolonepropionate hydrolase, UROC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131669 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction