Learn More
Peptides InternationalSupplier Diversity Partner Urokinase-Derived Peptide A6, 5mg, MF= C39H62N10O15 DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner UPA3038PI5MG
Ac-Lys-Pro-Ser-Ser-Pro-Pro-Glu-Glu-NH2; A6 Peptide; Urokinase Plasminogen Activator (136-143), human; Acetyl-Lysyl-Prolyl-Seryl-Seryl-Prolyl-Prolyl-Glutamyl-Glutamic Acid Amide; uPA-Derived Peptide A6, Application: CD44 Inhibitor; Anti-Tumor Properties, Notes: Sequence derived from peptide domain of human urokinase plasminogen activator (uPA). It is able to bind to and inhibit CD44 activity that contributes to tumorgenesis., salt: Trifluoroacetate Salt, one letter code: Ac-KPSSPPEE-NH2, MW: 910.99
Catalog No. 50-195-2444