Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Mouse anti-Human, Clone: UPK1A/2923, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30750210UG

 View more versions of this product

Catalog No. NB122477


Add to cart

Description

Description

Uroplakin Ia Monoclonal antibody specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

Uroplakin Ia
UPK1A/2923
Unconjugated
10 mM PBS with 0.05% BSA
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
11045
ELISA, Peptide Array
0.2 mg/ml
ELISA, Protein Array
Purified
Mouse
IgG2b Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit