Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ia Mouse anti-Human, Clone: UPK1A/2924, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30750320UG
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Protein Array.Specifications
Uroplakin Ia | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Protein Array | |
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
ELISA, Peptide Array | |
UPK1A/2924 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
11045 | |
Store at 4°C. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction