Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uroplakin Ib, Mouse anti-Human, DyLight 350, Clone: UPK1B/3081, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279933UV
Description
Uroplakin Ib Monoclonal antibody specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Uroplakin Ib | |
UPK1B/3081 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
UPK1B | |
Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title