Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ib, Mouse anti-Human, DyLight 488, Clone: UPK1B/3102, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279934G

Catalog No. NB012904


Add to cart

Description

Description

Uroplakin Ib Monoclonal antibody specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Uroplakin Ib
UPK1B/3102
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
UPK1B
Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit