Learn More
Biocare Medical, LLCSupplier Diversity Partner UROPLAKIN II 70TEST  DFS Item
Manufacturer:  Biocare Medical, LLCSupplier Diversity Partner OAI3051T60
Uroplakin II [BC21] is a highly specific antibody that may be useful in identifying tumors of urothelial origin. It has exhibited an increased sensitivity when compared to Uroplakin III [AU1] in urothelial bladder carcinoma. PATENT PENDING
Catalog No. 50-719-847