Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Uroplakin III Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119865
This item is not returnable.
View return policy
Description
Positive Control: Hela cell lysates, rat bladder tissue. Subcellular Location: Endoplasmic reticulum membrane.
UPK3A (Uroplakin 3A) is a Protein Coding gene. Diseases associated with UPK3A include Renal Hypodysplasia/Aplasia 3 and Ovarian Brenner Tumor. Among its related pathways are Bladder cancer. An important paralog of this gene is UPK3B.
Specifications
Uroplakin III | |
Polyclonal | |
Unconjugated | |
UPK3A | |
LOC105375355; p35; RGD1309615; UP3a; UP3b; UP3KA; UP3KB; UPIII; UPIIIA; UPIIIb; UPK3; Upk3a; UPK3A/B; Upk3b; uroplakin 3; uroplakin 3A; uroplakin 3B; uroplakin III; uroplakin IIIA; uroplakin IIIb; Uroplakin-3a; uroplakin-3b; urothelial differentiation-dependent marker | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105375355, 315190, 360790, 7380 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
O75631, Q9BT76 | |
UPK3A, UPK3B | |
Recombinant protein within human Uroplakin III aa 50-207. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction