Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner UROPORPHYRIN I 10 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner FH04010MG
Uroporphyrin I is stable for at least one year when stored as a solid, protected from moisture, at -20 °C. Protect from light.
Catalog No. 50-720-331