Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UROS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149131AP

Catalog No. 50-172-9829


Add to cart

Description

Description

The protein encoded by this gene catalyzes the fourth step of porphyrin biosynthesis in the heme biosynthetic pathway. Defects in this gene cause congenital erythropoietic porphyria (Gunther ft.s disease).
Specifications

Specifications

UROS
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10746, P51163
UROIIIS, uroporphyrinogen III synthase, UROS
Rabbit
IgG
150 μL
-20°C
Primary
22276, 309070, 7390
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
UROS
Liquid
Uros
UROS Fusion Protein Ag6711
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit