Learn More
Alfa Aesar™ Urotensin II, Goby
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66939MA
 View more versions of this product
Catalog No. AAJ66939MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1361.57
10mg
Ala-Gly-Thr-Ala-Asp-Cys-Phe-Trp-Lys-Tyr-Cys-Val, (Disulfide bridge: Cys6-Cys11
-30° to -10°C
HPLC
Solid
RUO
Urotensin II, goby