Learn More
Urotensin-II R/GPR14 Rat anti-Mouse, Alexa Fluor 488, Clone: 443707, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB9245G100UG
Catalog No. FAB9245G100
Add to cart
This item is not returnable. View return policy
Description
Description
Urotensin-II R/GPR14 Monoclonal antibody specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Urotensin-II R/GPR14
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rat
IgG2b
RUO
Primary
2837
Mouse
Flow Cytometry
443707
Flow Cytometry 0.25-1 ug/10^6 cells
G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UrotensinIIR, Urotensin-IIR, UTR, UTR2, UTS2R
Mouse Urotensin-II R synthetic peptide
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse Urotensin-II R. Stains mouse Urotensin-II R transfectants but not irrelevant transfectants in flow cytometry.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)