Learn More
UrotheliumMarker Mouse anti-Human, Janelia Fluor 549, Clone: LBS 8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250345JF549
Catalog No. NBP50345549
Add to cart
Description
Description
UrotheliumMarker Monoclonal antibody specifically detects UrotheliumMarker in Human samples. It is validated for Immunohistochemistry.
Specifications
Specifications
UrotheliumMarker
LBS 8
Immunohistochemistry
Purified
Human urothelial carcinoma -derived RT112 cells.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.1mL
Primary
Human