Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursodeoxycholic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
128-13-2
C24H40O4
392.58
MFCD00003680
RUDATBOHQWOJDD-UZVSRGJWSA-N
ursodeoxycholic acid,ursodiol,actigall,ursofalk,ursolvan,delursan,ursodeoxycholate,urso,urso forte,ursodesoxycholic acid
31401
CHEBI:9907
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C
128-13-2
C24H40O4
392.58
MFCD00003680
RUDATBOHQWOJDD-UZVSRGJWSA-N
ursodeoxycholic acid,ursodiol,actigall,ursofalk,ursolvan,delursan,ursodeoxycholate,urso,urso forte,ursodesoxycholic acid
31401
CHEBI:9907
(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
CC(CCC(=O)O)C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C
Packaging Glass bottle
Specific Rotation Condition +60.00 (20.00°C c=2,C2H5OH)
Quantity 1g
Specific Rotation +58° to +62° (20°C, 589nm) (c=2, ethanol)
Infrared Spectrum Authentic
Color Colorless
Melting Point 200°C to 206°C
Assay Percent Range 99%
Merck Index 15, 10074
Formula Weight 392.57
Physical Form Solution
Percent Purity ≥98.5% (Acid)
Chemical Name or Material Ursodeoxycholic acid
Show More Show Less