Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursodiol, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
128-13-2
C24H40O4
392.58
RUDATBOHQWOJDD-UZVSRGJWSA-N
(4R)-4-[(1R,3aS,3bR,4S,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
128-13-2
C24H40O4
392.58
RUDATBOHQWOJDD-UZVSRGJWSA-N
(4R)-4-[(1R,3aS,3bR,4S,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

U1304, 128-13-2, C24H40O4

Spectrum™ Ursodiol, USP is a bile acid used to prevent and dissolve certain types of gallstones. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 25 g
Melting Range 200-205°C
Specific Rotation +57° to +62°
Loss on Drying 0.005
CAS Max % 1
Percent Purity 98.5 to 101.5%
Grade USP