Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursolic acid, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Ursolic acid belongs to the class of triterpenoid compounds that occurs widely in medicinal herbs, food and other plants. It may possess pharmacological properties like hepta-protection, analgesic, cardiotonic, sedative and tonic effects.

$107.10 - $425.00

Specifications

Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Linear Formula C30H48O3
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5503
View Documents
MilliporeSigma Supelco
897975MGF
5 mg
Each for $107.10
Add to cart
 
11-101-5504
View Documents
MilliporeSigma Supelco
8979725MGF
25 mg
Each for $425.00
Add to cart
 
Description

Description

Food and Beverages
Specifications

Specifications

≥98.5% (HPLC)
Analytical Standard
C30H48O3
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit