Learn More
US Safety N95 Flat-Fold Disposable Respirator
Flat-fold N95 respirators fit conveniently in pockets for easy transporting.
Manufacturer:  US Safety AD4N95B240
 View more versions of this product
Catalog No. 19-825-554
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Adjustable nose clip — Integrated wire nosepiece with padded bridge for customized comfort
  • Single continuous strap — Provides equal upper and lower support, with no staples
  • Large breathing cavity — Unique shape allows easier, cooler breathing
  • Flat-fold convenience — Stores easily in a pocket for transportation
  • Individually packaged — Maintains product integrity and cleanliness
  • Optional valved design — For enhanced breathing comfort
  • Fits easily in a coat pocket; convenient dispenser box mounts on a shelf or wall, so you can put them right where they are needed most
Specifications
Specifications
N - Not Resistant to Oil
Flat-fold
N95
Universal
95%
AD4N95B240
Individually wrapped in polybags
Non-sterile
No
NIOSH 42 CFR 84, N95
Yes
General Purpose Particulate Respirator
No
Yes
Adjustable Nose Clip
20/Pk.
Single Continuous Strap