Learn More
US Safety N95 Flat-Fold Disposable Respirator
Flat-fold N95 respirators fit conveniently in pockets for easy transporting.
Specifications
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Filter Type N95
Filter Efficiency 95%
Design Type Flat-fold
View More Specs
Products 2
Catalog Number Mfr. No. With Exhalation Valve Price Quantity & Availability  
Catalog Number Mfr. No. With Exhalation Valve Price Quantity & Availability  
19-825-554
View Documents
US Safety
AD4N95B240
No This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-825-555
View Documents
US Safety
AD4N95VB120
Yes This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Adjustable nose clip — Integrated wire nosepiece with padded bridge for customized comfort
  • Single continuous strap — Provides equal upper and lower support, with no staples
  • Large breathing cavity — Unique shape allows easier, cooler breathing
  • Flat-fold convenience — Stores easily in a pocket for transportation
  • Individually packaged — Maintains product integrity and cleanliness
  • Optional valved design — For enhanced breathing comfort
  • Fits easily in a coat pocket; convenient dispenser box mounts on a shelf or wall, so you can put them right where they are needed most
Specifications
Specifications
NIOSH 42 CFR 84, N95
N95
95%
Flat-fold