Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier pH Storage Solution Products for Science Education

Used to replace spilled or contaminated solution.

Manufacturer:  Vernier PHSS

Catalog No. S16239ND


Add to cart

Description

Description

  • For use with: Go Wireless™ pH sensor, ORP Sensor, pH Sensor, Tris-Compatible Flat pH Sensor, Flat pH Electrode BNC, pH Electrode BNC
  • Quantity: 500mL bottle
Specifications

Specifications

pH Storage Solution
500 mL
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit