Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USF2 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for USF2

Manufacturer:  Abnova Corporation H00007392T01

Catalog No. 89-001-895


Add to cart

Description

Description

This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs.

  • Upstream transcription factor 2, c-fos interacting
  • Transfected cell line: 293T
  • Plasmid: pCMV-USF2 full-length
  • Host: Human
  • Theoretical molecular weight: 37kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

Specifications

Western Blot
293T
NM_003367.2
6.8
Transient overexpression cell Lysate was tested with Anti-USF2 antibody (H00007392-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
USF2 293T Cell Transient Overexpression Lysate(Denatured)
37
pCMV-USF2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit