Learn More
Aviva Systems Biology Corp USP12 Antibody - middle region (ARP55812_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55812_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human USP12. Alternate names: USP12L1, UBH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61622